Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 4.65 mm Dia, 2 in OAL
Catalog No. 5000536994
Encompass
This Class ZZ Minus Plug & Pin Gage is a precise measurement tool, featuring a 4.65 mm diameter and a 2 in overall length. It is constructed from steel and designed for applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A certificate of accuracy with NIST traceability is included.
  • Designed for precise measurement applications
  • Features a 4.65 mm diameter
  • Has a 2 in overall length
  • Constructed from durable steel
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices
Catalog No. 50-005-36994 Supplier MSC Supplier No. 40432106
This Class ZZ Minus Plug & Pin Gage is a precise measurement tool, featuring a 4.65 mm diameter and a 2 in overall length. It is constructed from steel and designed for applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A certificate of accuracy with NIST traceability is included.
  • Designed for precise measurement applications
  • Features a 4.65 mm diameter
  • Has a 2 in overall length
  • Constructed from durable steel
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices