Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 4.83 mm Dia, 2 in OAL, Steel
Catalog No. 5000516544
Encompass
This Class ZZ Minus Plug & Pin Gage is a precise measuring tool designed for accurate verification and inspection in various industrial and laboratory settings. With a diameter of 4.83 mm and an overall length of 2 in, this steel gage is ideal for economical general-purpose measurements, including checking hole sizes, gaging slot widths, and NoGo inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Provides a 10 microinch finish or better.
  • Gages over 0.0605 in are marked with size and direction of tolerance.
  • Individually packaged for protection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Offers added wear life compared to closer tolerance gages.
  • Economical solution for quick and accurate measurements.
Catalog No. 50-005-16544 Supplier MSC Supplier No. 40432288
This Class ZZ Minus Plug & Pin Gage is a precise measuring tool designed for accurate verification and inspection in various industrial and laboratory settings. With a diameter of 4.83 mm and an overall length of 2 in, this steel gage is ideal for economical general-purpose measurements, including checking hole sizes, gaging slot widths, and NoGo inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Provides a 10 microinch finish or better.
  • Gages over 0.0605 in are marked with size and direction of tolerance.
  • Individually packaged for protection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Offers added wear life compared to closer tolerance gages.
  • Economical solution for quick and accurate measurements.