Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 4.85 mm, 2 in OAL
Catalog No. 5000511598
Encompass
This Class ZZ Minus Plug & Pin Gage features a 4.85 mm diameter and a 2 inch overall length. Constructed from steel with a 10 micro coating/finish, it is suitable for various precision measurement tasks.
  • Steel construction
  • 10 micro coating/finish
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-11598 Supplier MSC Supplier No. 40432304
This Class ZZ Minus Plug & Pin Gage features a 4.85 mm diameter and a 2 inch overall length. Constructed from steel with a 10 micro coating/finish, it is suitable for various precision measurement tasks.
  • Steel construction
  • 10 micro coating/finish
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability