Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 5.75 mm, 2 in, Steel
Catalog No. 5000528912
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. It features a 5.75 mm diameter and a 2 inch overall length. With a 10 Micro finish and steel construction, it provides reliable performance. Key specifications include a tolerance of 0.00508 mm, making it suitable for various measuring applications.
  • Designed for accurate inspection
  • 5.75 mm diameter
  • 2 inch overall length
  • 10 Micro finish
  • Constructed from steel
  • Tolerance of 0.00508 mm
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
Catalog No. 50-005-28912 Supplier MSC Supplier No. 40433203
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. It features a 5.75 mm diameter and a 2 inch overall length. With a 10 Micro finish and steel construction, it provides reliable performance. Key specifications include a tolerance of 0.00508 mm, making it suitable for various measuring applications.
  • Designed for accurate inspection
  • 5.75 mm diameter
  • 2 inch overall length
  • 10 Micro finish
  • Constructed from steel
  • Tolerance of 0.00508 mm
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability