Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 6.05 mm, 2 in
Catalog No. 5000527859
Encompass
This Class ZZ Minus Plug & Pin Gage features a 6.05 mm diameter and a 2 inch overall length. Constructed from steel, it is designed for precise measurement applications with a tolerance of 0.00508 mm and a 10 Micro coating/finish.
  • Class ZZ Minus for precise measurements
  • 6.05 mm diameter
  • 2 in overall length
  • Made from steel
  • Features a 0.00508 mm tolerance
  • Includes a 10 Micro coating/finish
  • Comes with a Certificate of Accuracy with NIST Traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Can be used with verniers and other measuring devices
Catalog No. 50-005-27859 Supplier MSC Supplier No. 40433500
This Class ZZ Minus Plug & Pin Gage features a 6.05 mm diameter and a 2 inch overall length. Constructed from steel, it is designed for precise measurement applications with a tolerance of 0.00508 mm and a 10 Micro coating/finish.
  • Class ZZ Minus for precise measurements
  • 6.05 mm diameter
  • 2 in overall length
  • Made from steel
  • Features a 0.00508 mm tolerance
  • Includes a 10 Micro coating/finish
  • Comes with a Certificate of Accuracy with NIST Traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Can be used with verniers and other measuring devices