Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 6.38 mm Diameter, 2 inch OAL
Catalog No. 5000527159
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 6.38 mm diameter for precise measurement
  • 2 inch overall length
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Class ZZ Minus for specific gauging requirements
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Made in the United States
Catalog No. 50-005-27159 Supplier MSC Supplier No. 40433831
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 6.38 mm diameter for precise measurement
  • 2 inch overall length
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Class ZZ Minus for specific gauging requirements
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Made in the United States