Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 6.40 mm Dia, 2 inch OAL
Catalog No. 5000529110
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 6.40 mm diameter and 2-inch overall length, manufactured from durable steel. It is designed for accurate measurement and inspection tasks, commonly used for setting micrometers, NoGo inspection, and with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Precision ground to Class ZZ Minus tolerance.
  • Ideal for setting micrometers and other measuring instruments.
  • Suitable for NoGo inspection applications.
  • Constructed from high-quality steel for durability.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-29110 Supplier MSC Supplier No. 40433856
This Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 6.40 mm diameter and 2-inch overall length, manufactured from durable steel. It is designed for accurate measurement and inspection tasks, commonly used for setting micrometers, NoGo inspection, and with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Precision ground to Class ZZ Minus tolerance.
  • Ideal for setting micrometers and other measuring instruments.
  • Suitable for NoGo inspection applications.
  • Constructed from high-quality steel for durability.
  • Includes a certificate of accuracy with NIST traceability.