Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 7.26 mm, Steel
Catalog No. 5000528467
Encompass
This plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Constructed from durable steel, it features a 7.26 mm diameter and comes with a certificate of accuracy.
  • Can be used to set micrometers, for NoGo inspection, and with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Made from steel for durability
  • Overall length of 2 inches
  • Tolerance of 0.00508 mm for high precision
  • Features a 10 micro coating/finish
Catalog No. 50-005-28467 Supplier MSC Supplier No. 40434714
This plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Constructed from durable steel, it features a 7.26 mm diameter and comes with a certificate of accuracy.
  • Can be used to set micrometers, for NoGo inspection, and with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Made from steel for durability
  • Overall length of 2 inches
  • Tolerance of 0.00508 mm for high precision
  • Features a 10 micro coating/finish