Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 7.41 mm, Steel
Catalog No. 5000514065
Encompass
This Vermont Gage Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a 7.41 mm diameter, an overall length of two inches, and a Class ZZ Minus tolerance. Made from steel with a ten Micro finish, this gage is suitable for setting micrometers, NoGo inspection, and calibrating other measuring devices.
  • 7.41 mm diameter
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Made from steel
  • Features a 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices
Catalog No. 50-005-14065 Supplier MSC Supplier No. 40434862
This Vermont Gage Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a 7.41 mm diameter, an overall length of two inches, and a Class ZZ Minus tolerance. Made from steel with a ten Micro finish, this gage is suitable for setting micrometers, NoGo inspection, and calibrating other measuring devices.
  • 7.41 mm diameter
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Made from steel
  • Features a 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices