Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 7.95 mm, 2 in OAL
Catalog No. 5000528468
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage offers quick, accurate measurements for various gaging tasks, including hole diameters and Go/NoGo inspection. Featuring a 7.95 mm diameter and 2-inch overall length, this tool steel gage has a 10 microinch finish and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance of 0.00508 mm.
  • Made from tool steel with a 60/65 Rc hardness.
  • Features a 10 microinch or better finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for Go/NoGo inspection.
Catalog No. 50-005-28468 Supplier MSC Supplier No. 40435406
This Vermont Gage Class ZZ Minus Plug & Pin Gage offers quick, accurate measurements for various gaging tasks, including hole diameters and Go/NoGo inspection. Featuring a 7.95 mm diameter and 2-inch overall length, this tool steel gage has a 10 microinch finish and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance of 0.00508 mm.
  • Made from tool steel with a 60/65 Rc hardness.
  • Features a 10 microinch or better finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for Go/NoGo inspection.