Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 8.78 mm, 2 in
Catalog No. 5000533983
Encompass
This Class ZZ Minus plug and pin gage, with a diameter of 8.78 mm and an overall length of 2 in, is constructed from steel with a 10 micro coating/finish. It is designed for precise measurements and includes a Certificate of Accuracy with NIST Traceability. This gage is ideal for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspections.
  • Includes certificate of accuracy with NIST traceability.
  • Made of steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers.
  • Suitable for NoGo inspection.
  • Suitable for verniers and other measuring devices.
Catalog No. 50-005-33983 Supplier MSC Supplier No. 40436230
This Class ZZ Minus plug and pin gage, with a diameter of 8.78 mm and an overall length of 2 in, is constructed from steel with a 10 micro coating/finish. It is designed for precise measurements and includes a Certificate of Accuracy with NIST Traceability. This gage is ideal for setting micrometers, verniers, and other measuring devices, as well as for NoGo inspections.
  • Includes certificate of accuracy with NIST traceability.
  • Made of steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers.
  • Suitable for NoGo inspection.
  • Suitable for verniers and other measuring devices.