Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 9.00 mm, 2 in
Catalog No. 5000528421
Encompass
This plug and pin gage features a 9.00 mm diameter and a 2 inch overall length. Constructed from steel, it boasts a 10 Micro finish and includes a Certificate of Accuracy with NIST Traceability. It is designed for precise measurement applications such as setting micrometers and no-go inspection.
  • 9.00 mm measurement diameter
  • 2 inch overall length
  • Constructed from durable steel
  • Features a 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers and no-go inspection
  • Can be used with verniers and other measuring devices
Catalog No. 50-005-28421 Supplier MSC Supplier No. 40436453
This plug and pin gage features a 9.00 mm diameter and a 2 inch overall length. Constructed from steel, it boasts a 10 Micro finish and includes a Certificate of Accuracy with NIST Traceability. It is designed for precise measurement applications such as setting micrometers and no-go inspection.
  • 9.00 mm measurement diameter
  • 2 inch overall length
  • Constructed from durable steel
  • Features a 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers and no-go inspection
  • Can be used with verniers and other measuring devices