Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 9.68 mm Diameter, 2 Inch OAL
Catalog No. 5000527164
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. With a diameter of 9.68 mm and a 2-inch overall length, this gage is suitable for setting micrometers and for NoGo inspection, as well as use with verniers and other measuring devices. Crafted from durable steel and featuring a 10 Micro coating/finish, it is built for reliability and longevity. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring verifiable precision for critical applications.
  • Precision instrument for accurate measurement and inspection.
  • Suitable for setting micrometers, NoGo inspection, and use with other measuring devices.
  • Made from durable steel for reliability and longevity.
  • Features a 10 Micro coating/finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
Catalog No. 50-005-27164 Supplier MSC Supplier No. 40437139
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. With a diameter of 9.68 mm and a 2-inch overall length, this gage is suitable for setting micrometers and for NoGo inspection, as well as use with verniers and other measuring devices. Crafted from durable steel and featuring a 10 Micro coating/finish, it is built for reliability and longevity. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring verifiable precision for critical applications.
  • Precision instrument for accurate measurement and inspection.
  • Suitable for setting micrometers, NoGo inspection, and use with other measuring devices.
  • Made from durable steel for reliability and longevity.
  • Features a 10 Micro coating/finish.
  • Includes a Certificate of Accuracy with NIST Traceability.