Learn More
MSC Vermont Gage Plug & Pin Gage, ZZ Minus, 0.6660 in Dia, 2 in OAL
Catalog No. 5000536912
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. Made from durable steel, it features a 0.6660 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, performing no-go inspections, and calibrating verniers and other measuring devices.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers.
  • Suitable for no-go inspection.
  • Can be used with verniers and other measuring devices.
  • Constructed from steel.
  • Provides a tolerance of 0.000200 "
Catalog No. 50-005-36912 Supplier MSC Supplier No. 89066609
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. Made from durable steel, it features a 0.6660 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, performing no-go inspections, and calibrating verniers and other measuring devices.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers.
  • Suitable for no-go inspection.
  • Can be used with verniers and other measuring devices.
  • Constructed from steel.
  • Provides a tolerance of 0.000200 "