Learn More
MSC Vermont Gage Plus Plug Gage, 19.03 mm, Class ZZ, 0.00508 mm
Catalog No. 5000527968
Encompass
This Vermont Gage Class ZZ Plus Plug Gage features a 19.03 mm diameter and a 0.00508 mm tolerance. Constructed from durable tool steel, it is designed for accurate go inspection and setting various measuring devices such as micrometers and verniers.
  • For go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Made from tool steel.
  • 10 micro finish.
  • 2.0000 in overall length.
Catalog No. 50-005-27968 Supplier MSC Supplier No. 68110089
This Vermont Gage Class ZZ Plus Plug Gage features a 19.03 mm diameter and a 0.00508 mm tolerance. Constructed from durable tool steel, it is designed for accurate go inspection and setting various measuring devices such as micrometers and verniers.
  • For go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Made from tool steel.
  • 10 micro finish.
  • 2.0000 in overall length.