Learn More
MSC Vermont Gage Replacement Pin, 0.7400 in, Class ZZ Minus, Steel
Catalog No. 5000515936
Encompass
This Class ZZ Minus replacement pin gage is an economical general-purpose tool designed for quick and accurate measurements. It is suitable for checking hole sizes, calculating distances between holes, gaging slot widths, and checking locations, as well as for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Class ZZ tolerance (.0002 in or .005 mm).
  • NoGo (minus) tolerance type.
  • Made from tool steel with 60/65 Rc hardness.
  • Two inch overall length.
  • Features a 10 microinch finish or better.
  • Round within .0001 in or .0025 mm.
  • NIST traceable for accuracy.
  • Marked with size and direction of tolerance (for gages over .0605 in).
  • Certificate of accuracy included.
  • Individually packaged.
Catalog No. 50-005-15936 Supplier MSC Supplier No. 89074009
This Class ZZ Minus replacement pin gage is an economical general-purpose tool designed for quick and accurate measurements. It is suitable for checking hole sizes, calculating distances between holes, gaging slot widths, and checking locations, as well as for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Class ZZ tolerance (.0002 in or .005 mm).
  • NoGo (minus) tolerance type.
  • Made from tool steel with 60/65 Rc hardness.
  • Two inch overall length.
  • Features a 10 microinch finish or better.
  • Round within .0001 in or .0025 mm.
  • NIST traceable for accuracy.
  • Marked with size and direction of tolerance (for gages over .0605 in).
  • Certificate of accuracy included.
  • Individually packaged.