This Steel Class ZZ Gage Pin is designed for precise measurement applications, offering a diameter of .3975 inches and a tight Class ZZ tolerance for accurate gaging. Constructed from durable tool steel, it is suitable for various industrial and laboratory settings requiring high precision.
- Class ZZ tolerance for precise measurements
- Available in Go (Plus) or NoGo (Minus) configurations
- Diameters available in .0005 inch or 0.01 mm increments
- Set increments of .001 inch or 0.02 mm
- Made from tool steel with 60/65 Rc hardness
- 2 inch length for versatile use
- Surface finish of 10 microinch or better
- Round within .0001 inch or 0.0025 mm
- Gages over .0605 inch are marked with diameter and direction of tolerance
- Includes a certificate of accuracy
- Calibration certificates, ISO 17025 accredited and non-accredited, may be purchased