Learn More
MSC Vermont Gage Steel Plug Gage, Class ZZ, 0.6880 Dia, Plus Tolerance
Catalog No. 5000515076
Encompass
This Vermont Gage Steel Plug Gage, Class ZZ, features a 0.6880 inch diameter and plus tolerance. It is designed for quick and accurate measurements, including hole diameters, slot widths, and for setting micrometers, verniers, and other measuring devices.
  • Quick and accurate measurements
  • Measures hole diameters, slot widths, and locations
  • Used for Go/NoGo inspection
  • Sets micrometers, verniers, and other measuring devices
  • Marked with diameter and tolerance direction on gages over 0.0605 in
  • Includes certificate of accuracy
  • Optional ISO 17025 accredited and non-accredited calibration certificates
  • Class ZZ tolerance of 0.0002 in or 0.005 mm
  • Go (plus) or NoGo (minus) configuration
  • Constructed from tool steel, 60/65 Rc hardness
  • 2 in long with 10 microinch finish or better
  • Round within 0.0001 in or 0.0025 mm
Catalog No. 50-005-15076 Supplier MSC Supplier No. 88968805
This Vermont Gage Steel Plug Gage, Class ZZ, features a 0.6880 inch diameter and plus tolerance. It is designed for quick and accurate measurements, including hole diameters, slot widths, and for setting micrometers, verniers, and other measuring devices.
  • Quick and accurate measurements
  • Measures hole diameters, slot widths, and locations
  • Used for Go/NoGo inspection
  • Sets micrometers, verniers, and other measuring devices
  • Marked with diameter and tolerance direction on gages over 0.0605 in
  • Includes certificate of accuracy
  • Optional ISO 17025 accredited and non-accredited calibration certificates
  • Class ZZ tolerance of 0.0002 in or 0.005 mm
  • Go (plus) or NoGo (minus) configuration
  • Constructed from tool steel, 60/65 Rc hardness
  • 2 in long with 10 microinch finish or better
  • Round within 0.0001 in or 0.0025 mm