Learn More
MSC Vermont Gage Threaded Ring Gage, #10-32 Thread, UNF, Class 2A, Go
Catalog No. 5000529209
Encompass
This threaded ring gage is designed for precise measurement applications. It features a #10-32 UNF thread with a Class 2A classification, indicating it is a 'Go' gage for verifying external threads.
  • Functions as a 'Go' gage for external thread verification.
  • Thread size is #10-32.
  • UNF thread type.
  • Class 2A classification.
  • Single ring design.
  • Constructed from durable tool steel.
  • Conforms to NIST standards.
  • Manufactured in the United States.
  • Includes a certificate of compliance.
Catalog No. 50-005-29209 Supplier MSC Supplier No. 05418322
This threaded ring gage is designed for precise measurement applications. It features a #10-32 UNF thread with a Class 2A classification, indicating it is a 'Go' gage for verifying external threads.
  • Functions as a 'Go' gage for external thread verification.
  • Thread size is #10-32.
  • UNF thread type.
  • Class 2A classification.
  • Single ring design.
  • Constructed from durable tool steel.
  • Conforms to NIST standards.
  • Manufactured in the United States.
  • Includes a certificate of compliance.