Learn More
MSC Vermont Gage Threaded Ring Gage, 3/4-16 Thread, UNF, Class 3A, Go
Catalog No. 5000516887
Encompass
This threaded ring gage from Vermont Gage is designed to inspect external screw threads for accuracy. It features a 3/4-16 UNF thread size with a Class 3A fit, indicating precise manufacturing standards. Made from durable tool steel, this "Go" gage ensures that components meet dimensional requirements for proper fit.
  • Inspects external screw threads for accuracy.
  • Features a 3/4-16 UNF thread size.
  • Ensures a class 3A fit.
  • Made from durable tool steel.
  • Conforms to NIST standards.
  • Single ring design for specific inspections.
Catalog No. 50-005-16887 Supplier MSC Supplier No. 05447164
This threaded ring gage from Vermont Gage is designed to inspect external screw threads for accuracy. It features a 3/4-16 UNF thread size with a Class 3A fit, indicating precise manufacturing standards. Made from durable tool steel, this "Go" gage ensures that components meet dimensional requirements for proper fit.
  • Inspects external screw threads for accuracy.
  • Features a 3/4-16 UNF thread size.
  • Ensures a class 3A fit.
  • Made from durable tool steel.
  • Conforms to NIST standards.
  • Single ring design for specific inspections.