Learn More
MSC Vermont Gage Threaded Ring Gage, #3-56 UNF, Class 2A, No Go
Catalog No. 5000533392
Encompass
This Threaded Ring Gage is designed for precision measurement. It features a #3-56 UNF thread with a Class 2A tolerance, specifically a No Go type, ensuring accurate checks for external threads.
  • Designed as a "no go" gage for checking thread limits.
  • Features a #3-56 UNF thread size.
  • Class 2A classification ensures precise measurements.
  • Constructed from durable tool steel.
  • Single ring design for specific applications.
  • Manufactured in the United States.
Catalog No. 50-005-33392 Supplier MSC Supplier No. 05413570
This Threaded Ring Gage is designed for precision measurement. It features a #3-56 UNF thread with a Class 2A tolerance, specifically a No Go type, ensuring accurate checks for external threads.
  • Designed as a "no go" gage for checking thread limits.
  • Features a #3-56 UNF thread size.
  • Class 2A classification ensures precise measurements.
  • Constructed from durable tool steel.
  • Single ring design for specific applications.
  • Manufactured in the United States.