Learn More
MSC Vermont Gage Threaded Ring Gage, 3/8-16 UNC, Class 2A, No Go
Catalog No. 5000529994
Encompass
This threaded ring gage is designed for inspecting 3/8-16 UNC threads. It features a Class 2A classification and is a 'No Go' gage, ensuring precise measurement of thread tolerance. Constructed from durable tool steel, it meets NIST standards for accuracy.
  • No Go functionality for precise thread inspection.
  • Designed for 3/8-16 thread size.
  • Compatible with UNC thread types.
  • Features a Class 2A classification.
  • Single ring design.
  • Constructed from tool steel for durability.
  • Meets NIST standards for reliability.
  • Manufactured in the United States.
Catalog No. 50-005-29994 Supplier MSC Supplier No. 05422175
This threaded ring gage is designed for inspecting 3/8-16 UNC threads. It features a Class 2A classification and is a 'No Go' gage, ensuring precise measurement of thread tolerance. Constructed from durable tool steel, it meets NIST standards for accuracy.
  • No Go functionality for precise thread inspection.
  • Designed for 3/8-16 thread size.
  • Compatible with UNC thread types.
  • Features a Class 2A classification.
  • Single ring design.
  • Constructed from tool steel for durability.
  • Meets NIST standards for reliability.
  • Manufactured in the United States.