Learn More
MSC Vermont Gage Threaded Ring Gage, 5/16-24, UNF, Class 2A, No Go
Catalog No. 5000516860
Encompass
The Threaded Ring Gage is designed for inspecting externally threaded parts. This specific model is a No Go gage with a 5/16-24 UNF thread, Class 2A, and is constructed from durable tool steel, making it suitable for verifying the maximum pitch diameter of threads.
  • Inspects externally threaded parts
  • Verifies maximum pitch diameter
  • Made of durable tool steel
  • Adjustable design
  • Includes certificate of accuracy traceable to NIST
  • Uncoated finish
  • Hardness of RC 60-62
Catalog No. 50-005-16860 Supplier MSC Supplier No. 05421250
The Threaded Ring Gage is designed for inspecting externally threaded parts. This specific model is a No Go gage with a 5/16-24 UNF thread, Class 2A, and is constructed from durable tool steel, making it suitable for verifying the maximum pitch diameter of threads.
  • Inspects externally threaded parts
  • Verifies maximum pitch diameter
  • Made of durable tool steel
  • Adjustable design
  • Includes certificate of accuracy traceable to NIST
  • Uncoated finish
  • Hardness of RC 60-62