Learn More
MSC Vermont Gage Threaded Ring Gage, #5-40 Thread, UNC, Class 2A, No Go
Catalog No. 5000511275
Encompass
This threaded ring gage is designed for inspecting #5-40 UNC threads. It is a Class 2A 'No Go' gage, manufactured from durable tool steel, ensuring precision and reliability for quality control applications.
  • No go functionality for precise thread measurement.
  • Designed for #5-40 thread size.
  • Supports UNC thread type.
  • Features 2A classification.
  • Single ring design.
  • Constructed from durable tool steel.
  • Complies with NIST standards.
  • Manufactured in the United States.
Catalog No. 50-005-11275 Supplier MSC Supplier No. 05415419
This threaded ring gage is designed for inspecting #5-40 UNC threads. It is a Class 2A 'No Go' gage, manufactured from durable tool steel, ensuring precision and reliability for quality control applications.
  • No go functionality for precise thread measurement.
  • Designed for #5-40 thread size.
  • Supports UNC thread type.
  • Features 2A classification.
  • Single ring design.
  • Constructed from durable tool steel.
  • Complies with NIST standards.
  • Manufactured in the United States.