Learn More
MSC Vermont Gage Threaded Ring Gage, M6 x 1.00, Class 6G, No Go
Catalog No. 5000532938
Encompass
This threaded ring gage is designed for M6 x 1.00 metric threads. It functions as a Class 6G No Go gage, used to verify the maximum material limit of a threaded component. The gage is constructed from tool steel and comes with NIST traceability certification.
  • Designed for M6 x 1.00 metric threads.
  • Verifies maximum material limit of threaded components.
  • Constructed from tool steel.
  • Includes NIST traceability certification.
Catalog No. 50-005-32938 Supplier MSC Supplier No. 05346614
This threaded ring gage is designed for M6 x 1.00 metric threads. It functions as a Class 6G No Go gage, used to verify the maximum material limit of a threaded component. The gage is constructed from tool steel and comes with NIST traceability certification.
  • Designed for M6 x 1.00 metric threads.
  • Verifies maximum material limit of threaded components.
  • Constructed from tool steel.
  • Includes NIST traceability certification.