Learn More
MSC Vermont Gage Threaded Ring Gage, No Go, #4-40, UNC, Class 2A
Catalog No. 5000529997
Encompass
This product is a threaded ring gage designed for inspecting #4-40 UNC 2A threads, featuring a "No Go" configuration for precise measurement. It is constructed from durable tool steel, ensuring reliability and accuracy in various applications.
  • No go configuration for precise thread inspection.
  • Designed for #4-40 UNC threads.
  • Class 2A accuracy for consistent results.
  • Single ring design.
  • Constructed from tool steel for durability.
  • Manufactured in the United States.
  • Meets NIST 41111619 standards.
Catalog No. 50-005-29997 Supplier MSC Supplier No. 05414412
This product is a threaded ring gage designed for inspecting #4-40 UNC 2A threads, featuring a "No Go" configuration for precise measurement. It is constructed from durable tool steel, ensuring reliability and accuracy in various applications.
  • No go configuration for precise thread inspection.
  • Designed for #4-40 UNC threads.
  • Class 2A accuracy for consistent results.
  • Single ring design.
  • Constructed from tool steel for durability.
  • Manufactured in the United States.
  • Meets NIST 41111619 standards.