Learn More
MSC Vermont Gage Trilock Plug Gage, 1.3083 in, Class ZZ Plus
Catalog No. 5000531169
Encompass
This Trilock plug gage is designed for precise measurements in various applications, offering an economical solution for verifying hole sizes, distances, and slot widths. It features a Class ZZ Plus tolerance for accuracy and provides added wear life for extended use.
  • Features a Class ZZ tolerance of .0002 in
  • Manufactured from tool steel with 60/65 Rc hardness
  • NIST traceable for quality assurance
  • Length of 2 in
  • Surface finish of 10 microinch or better
  • Roundness within .0001 in
  • Marked with size and direction of tolerance for easy identification
  • Includes a certificate of accuracy
Catalog No. 50-005-31169 Supplier MSC Supplier No. 64759947
This Trilock plug gage is designed for precise measurements in various applications, offering an economical solution for verifying hole sizes, distances, and slot widths. It features a Class ZZ Plus tolerance for accuracy and provides added wear life for extended use.
  • Features a Class ZZ tolerance of .0002 in
  • Manufactured from tool steel with 60/65 Rc hardness
  • NIST traceable for quality assurance
  • Length of 2 in
  • Surface finish of 10 microinch or better
  • Roundness within .0001 in
  • Marked with size and direction of tolerance for easy identification
  • Includes a certificate of accuracy