Learn More
MSC Vermont Gage Trilock Plug Gage, 1.5000 Plus, Class ZZ, Tool Steel
Catalog No. 5000513849
Encompass
This Vermont Gage Trilock Plug Gage, with a size of 1.5000 in, is a Class ZZ gage designed for inspecting large diameters. It provides quick and accurate measurements for various gaging applications, including hole sizes, slot widths, and Go/NoGo checks.
  • Economical, general-purpose gage for quick and accurate measurements
  • Suitable for measuring hole sizes, slot widths, and locations
  • Used for Go/NoGo gaging
  • Class ZZ tolerance of ±.0002 in (±.005 mm)
  • NIST traceable
  • Round within .0001 in (.0025 mm)
  • 10 microinch finish or better
  • Marked with size and tolerance direction
  • Certificate of accuracy included
  • Constructed from tool steel for durability
Catalog No. 50-005-13849 Supplier MSC Supplier No. 64758626
This Vermont Gage Trilock Plug Gage, with a size of 1.5000 in, is a Class ZZ gage designed for inspecting large diameters. It provides quick and accurate measurements for various gaging applications, including hole sizes, slot widths, and Go/NoGo checks.
  • Economical, general-purpose gage for quick and accurate measurements
  • Suitable for measuring hole sizes, slot widths, and locations
  • Used for Go/NoGo gaging
  • Class ZZ tolerance of ±.0002 in (±.005 mm)
  • NIST traceable
  • Round within .0001 in (.0025 mm)
  • 10 microinch finish or better
  • Marked with size and tolerance direction
  • Certificate of accuracy included
  • Constructed from tool steel for durability