Learn More
MSC Vermont Gage Pin Gage, Tool Steel, Class ZZ, 0.7135 in., 1 ea.
Catalog No. 5000514155
Encompass
Vermont Gage Class ZZ pin gages provide accurate and efficient measurement of hole diameters, slot widths, and locations. These tools are suitable for use as Go/NoGo inspection gages, as well as for setting micrometers, verniers, and other precision measuring instruments.
  • Manufactured from tool steel with a hardness of 60 to 65 Rc
  • Diameter of 0.7135 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Surface finish of 10 microinch
  • Go (Plus) type gage
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-14155 Supplier MSC Supplier No. 07509292
Vermont Gage Class ZZ pin gages provide accurate and efficient measurement of hole diameters, slot widths, and locations. These tools are suitable for use as Go/NoGo inspection gages, as well as for setting micrometers, verniers, and other precision measuring instruments.
  • Manufactured from tool steel with a hardness of 60 to 65 Rc
  • Diameter of 0.7135 in. with a tolerance of 0.0002 in.
  • Overall length of 2 in.
  • Surface finish of 10 microinch
  • Go (Plus) type gage
  • Includes certificate of accuracy with NIST traceability