Learn More
Medchemexpress LLC Vernakalant hydrochloride | 748810-28-8 | MFCD09833303 | ≥98.0% | 385.93 g·mol⁻¹ | C20H32ClNO4 | 5 MG
SDP
Catalog No. 5000221654
Encompass_Preferred
Vernakalant hydrochloride is a research-grade antiarrhythmic compound that acts as a mixed voltage- and frequency-dependent sodium channel blocker and an atria-preferred potassium channel blocker. It is used in electrophysiology and cardiac ion-channel studies to investigate mechanisms of atrial fibrillation and to evaluate potential therapeutic effects. Consult the manufacturer datasheet and SDS for purity, handling, and storage details.
  • Mixed voltage- and frequency-dependent sodium channel blocker and atria-preferred potassium channel blocker.
  • Useful for studies of atrial fibrillation and cardiac electrophysiology.
  • Suitable for in vitro pharmacology and ion-channel assays.
  • Supplied in small research quantities for preclinical experiments.
  • Refer to the product datasheet and safety data sheet for purity, storage, and handling information.
Catalog No. 50-002-21654 Supplier Medchemexpress LLC Supplier No. HY141835MG
Vernakalant hydrochloride is a research-grade antiarrhythmic compound that acts as a mixed voltage- and frequency-dependent sodium channel blocker and an atria-preferred potassium channel blocker. It is used in electrophysiology and cardiac ion-channel studies to investigate mechanisms of atrial fibrillation and to evaluate potential therapeutic effects. Consult the manufacturer datasheet and SDS for purity, handling, and storage details.
  • Mixed voltage- and frequency-dependent sodium channel blocker and atria-preferred potassium channel blocker.
  • Useful for studies of atrial fibrillation and cardiac electrophysiology.
  • Suitable for in vitro pharmacology and ion-channel assays.
  • Supplied in small research quantities for preclinical experiments.
  • Refer to the product datasheet and safety data sheet for purity, storage, and handling information.