Learn More
Medchemexpress LLC Vernakalant hydrochloride | 748810-28-8 | MFCD09833303 | 99.3% | C20H32ClNO4 | 10MG
SDP
Catalog No. 5000258919
Encompass_Preferred
Vernakalant hydrochloride is a research chemical that acts as a mixed voltage- and frequency-dependent sodium channel blocker and an atria-preferred potassium channel blocker, used to investigate atrial electrophysiology and arrhythmia mechanisms. It is supplied in solid and DMSO solution formats and is characterized by defined molecular properties for in vitro studies.
  • High purity (99.33%).
  • Available as solid and 10 mM DMSO solution.
  • Suitable for in vitro electrophysiology and pharmacology studies.
  • Molecular formula C20H32ClNO4; molecular weight 385.93 g·mol⁻¹.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-58919 Supplier Medchemexpress LLC Supplier No. HY1418310MG
Vernakalant hydrochloride is a research chemical that acts as a mixed voltage- and frequency-dependent sodium channel blocker and an atria-preferred potassium channel blocker, used to investigate atrial electrophysiology and arrhythmia mechanisms. It is supplied in solid and DMSO solution formats and is characterized by defined molecular properties for in vitro studies.
  • High purity (99.33%).
  • Available as solid and 10 mM DMSO solution.
  • Suitable for in vitro electrophysiology and pharmacology studies.
  • Molecular formula C20H32ClNO4; molecular weight 385.93 g·mol⁻¹.
  • For research use only; not for human or veterinary use.