Learn More
Selleck Chemical LLC Vernakalant Hydrochloride
SDP
Catalog No. 503137150
Encompass_Preferred
Vernakalant (RSD-1235 MK-6621) is a novel frequency-dependent Na channel and early activating K channel blocker that selectively prolongs the atrial refractory period
Catalog No. 50-313-7150 Supplier Selleck Chemical LLC Supplier No. E01445MG
Vernakalant (RSD-1235 MK-6621) is a novel frequency-dependent Na channel and early activating K channel blocker that selectively prolongs the atrial refractory period