Learn More
Vernier Vernier Air Spectrum Tube
SDPSCI_ED
Catalog No. S16256ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Air Spectrum Tube
Catalog No. Product Type
S16256ND Air Spectrum Tube
1 options
Catalog No. S16256ND Supplier Vernier Supplier No. STAIR
Add to Cart
Add to Cart
Gas tubes for qualitative study by students noting more spectral lines and unique characteristic lines. For use with the Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply.
  • Permanently enclosed in plastic carriers to prevent breakage
  • No through-the-glass electrodes
  • Ultrasafe
  • Long lasting and superior to older designs
  • For grades 9 and up
Specifications
For Use With (Equipment) Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply
Material Plastic
Product Type Air Spectrum Tube
WARNING: Cancer - www.P65Warnings.ca.gov