Learn More
Vernier Blood Pressure Sensor
SDPSCI_ED
Catalog No. S16101ND Shop All Vernier Software Products
Change view
Click to view available options
Size:
Adult
Catalog No. Size
S16101ND Adult
1 options
This item is intended for educational purposes only.
Catalog No. S16101ND Supplier Vernier Supplier No. BPSBTA
Add to Cart
Add to Cart
This item is intended for educational purposes only.
Includes
Blood Pressure Sensor, Standard adult size adjustable cuff (27 to 39cm), Bulb pump (with release valve), User manual
Non-invasive sensor designed to measure human blood pressure.
  • Measures systolic, diastolic and mean arterial pressure utilizing oscillometric technique
  • Reports pulse rate
  • Compatible with LabQuest™ 2, Original LabQuest, LabQuest Mini, Go!Link™, SensorDAQ™, LabPro™, EasyLink™, TI-Nspire Lab Cradle™ interfaces
  • Recommended for College and High School
  • Requires compatible interface and software to collect data (sold separately)
  • Sensing element: SSCMRRN005PGAA5
  • Combined linearity and hysteresis: typical ±0.25%
Note: For replacement cuffs, see Vernier Blood Pressure Sensor Cuffs (CUFFLG, CUFFSM, CUFFSTD).
Specifications
Size Adult
Includes Blood Pressure Sensor, Standard adult size adjustable cuff (27 to 39cm), Bulb pump (with release valve), User manual