Learn More
Vernier BlueView Transilluminator
SDPSCI_ED
Catalog No. S16100ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
BlueView Transilluminator
Catalog No. Product Type
S16100ND BlueView Transilluminator
1 options
This item is intended for educational purposes only.
Catalog No. S16100ND Supplier Vernier Supplier No. BLUEVIEW
Add to Cart
Add to Cart
This item is intended for educational purposes only.
Uses super bright blue LEDs to illuminate electrophoresis gels stained with SYBR Safe stain.
Specifications
Product Type BlueView Transilluminator
WARNING: Cancer - www.P65Warnings.ca.gov