Learn More
Surgical Design™ Vernier Caliper
Catalog No. 21124917 Shop All Surgical Design Products
Catalog No. 21-124-917 Supplier Surgical Design™ Supplier No. MT001
Add to Cart
Add to Cart
Vernier caliper provides accurate internal, external, and depth measurements for laboratory, industrial, educational, and diagnostic applications.
  • High-precision scale for accurate internal, external, and depth measurements
  • Smooth-gliding vernier mechanism ensures stable, repeatable readings
  • Durable stainless-steel construction resistant to wear, corrosion, and lab chemicals
  • Clear, easy-to-read vernier markings for precise interpretation
  • Locking screw feature maintains measurements during use
  • Ideal for lab, workshop, engineering, and educational applications