Learn More
Electron Microscopy Sciences Vernier Caliper, Double scale, Nickel Plated
SDP
Catalog No. 50996490
Encompass_Preferred
Double scale vernier caliper. Nickel Plated Scale. Inch Scale: Reading from 0 to 5" in 1/16" divisions. Metric Scale: Reading from 0 to 120 mm in 1 mm divisions. Readable on the vernier scale to 1/128" and/or to 0.1 mm. Has a roller adjustment and it is packed in a plastic case.
Catalog No. 50-996-490 Supplier Electron Microscopy Sciences Supplier No. 62064
Add to Cart
Add to Cart
Double scale vernier caliper. Nickel Plated Scale. Inch Scale: Reading from 0 to 5" in 1/16" divisions. Metric Scale: Reading from 0 to 120 mm in 1 mm divisions. Readable on the vernier scale to 1/128" and/or to 0.1 mm. Has a roller adjustment and it is packed in a plastic case.