Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Class 2 Laser Pointer
SDPSCI_ED

Catalog No. s16182nd Shop All Vernier Software Products
Change view
Click to view available options
Class 2 Laser Pointer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16182ND Class 2 Laser Pointer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16182ND Supplier Vernier Supplier No. LASER
Only null left
Add to Cart
Add to Cart

For use with Photogate (S16279ND).

  • Class 2 (>1mW) laser pointer can be used with Vernier Photogate to produce gate that is wider than normal 7.5cm spacing
  • Measures speed of automobile or of person running
  • Laser case is made of aluminum, and it uses two AAA batteries (included)
  • Wavelength of light is 650nm
  • Momentary switch (normally off) controls laser beam

Specifications

Product Type Class 2 Laser Pointer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.