Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Conductivity Standard Solution (Low)
SDPSCI_ED

Catalog No. s16130nd Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Conductivity probe
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S16130ND Conductivity probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16130ND Supplier Vernier Supplier No. CONLST
Only null left
Add to Cart
Add to Cart

Calibrate conductivity probe in the high range (0 to 200μS/cm)

  • Concentration: 150μS/cm
  • Available in 500mL bottle

Specifications

For Use With (Equipment) Conductivity probe
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.