Learn More
Vernier Conductivity Standard Solution (Middle)
SDPSCI_ED
Catalog No. S16131ND Shop All Vernier Software Products
Catalog No. S16131ND Supplier Vernier Supplier No. CONMST
Add to Cart
Add to Cart
Calibrate conductivity probe in the high range (0 to 2000Glc muS/cm)
  • Concentration: 1413μS/cm
  • Available in 500mL bottle
WARNING: Cancer - www.P65Warnings.ca.gov