Learn More
Grainger Vernier Depth Gage 0-300mm
Catalog No. 502765740
Encompass_Preferred
The Grainger website features a variety of precision depth micrometers and depth gauges for sale. These tools are designed for accurate inside, outside, and depth measurements. They feature precision ground, hardened and lapped measuring rods. Resolution can be as fine as 0.0001" (0.001mm). Common measuring ranges are 0-1" to 0-6". These high quality tools are made by brands like Mitutoyo, Starrett, Fowler and more. They feature ratchet stop mechanisms and carbide measuring faces. Many include protective cases. These precision measuring tools are essential for machinists, metalworkers, engineers and quality control professionals.
Catalog No. 50-276-5740 Supplier Grainger Supplier No. 16X248
The Grainger website features a variety of precision depth micrometers and depth gauges for sale. These tools are designed for accurate inside, outside, and depth measurements. They feature precision ground, hardened and lapped measuring rods. Resolution can be as fine as 0.0001" (0.001mm). Common measuring ranges are 0-1" to 0-6". These high quality tools are made by brands like Mitutoyo, Starrett, Fowler and more. They feature ratchet stop mechanisms and carbide measuring faces. Many include protective cases. These precision measuring tools are essential for machinists, metalworkers, engineers and quality control professionals.