Learn More
Vernier Drop Counter
SDPSCI_ED
Catalog No. S16274ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Digital Drop Counter
Catalog No. Product Type
S16274ND Digital Drop Counter
1 options
Catalog No. S16274ND Supplier Vernier Supplier No. VDCBTD
Add to Cart
Add to Cart
Digital sensor used for performing accurate, automatic titrations.
Precisely records number of drops of titrant added during titration, which is automatically converted into volume.
  • Adjustable clamp, fits most laboratory ring stands and lattices
  • Wide drop-detecting area
  • Large sensor slot, fits most conventional stick-style sensors
  • Smaller, adjustable slot for temperature sensors
  • Accurate drop counting at rates up to 6 drops/second
  • Red LED, near the detecting area, flashes to indicate a drop has been recorded
  • Can be used in conjunction with sensors such as pH or ORP sensor or conductivity probe
  • Recommended for College and High School
Specifications
Product Type Digital Drop Counter
For Use With (Application) Titrations
Includes Plastic reagent reservoir, plastic valve with a double stopcock fitting, Vernier microstirrer (S16012ND) and user manual