Learn More
Vernier Vernier Flow Head
SDPSCI_ED
Catalog No. S16253ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Flow Head
Catalog No. Product Type
S16253ND Flow Head
1 options
This item is intended for educational purposes only.
Catalog No. S16253ND Supplier Vernier Supplier No. SPRFLOW
Add to Cart
Add to Cart
This item is intended for educational purposes only.
For use with Vernier Spirometer.
  • Extra flow head eliminates the need for equipment downtime while sterilizing your original flow head
  • Consists of a polycarbonate outer body with a stainless-steel inner mesh
Specifications
Product Type Flow Head
WARNING: Cancer - www.P65Warnings.ca.gov