Learn More
Description
- Sold as pack of 4 septa
- Color: green
Specifications
Specifications
| For Use With (Equipment) | Vernier Mini GC Gas Chromatograph |
| Product Type | GC Septa |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

