Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier KidWind Advanced Wind Experiment Kit Nacelle
SDPSCI_ED

Catalog No. S04800ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Kit, Advanced Wind Experiment, Nacelle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S04800ND Kit, Advanced Wind Experiment, Nacelle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04800ND Supplier Vernier Supplier No. KWAWXN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Gear set with spool, KidWind windturbine generator with wires, High torque generator with wires, KidWind Dowels, Drivertrain set

Enables number of advanced experiments, especially with GENPack generator module.

  • Contains unassembled parts of Advanced Wind Turbine Nacelle
  • Students can build their own wind turbine and experiment with wind power
  • Try different gear ratios, blade designs, generators and more in creative, hands-on inquiry into wind energy science
  • Also converts for conducting weightlifting experiments, making it great option for studying mechanical and electrical energy
  • Once assembled, nacelle mounts on wind turbine tower or on standard 1 in. PVC pipe

Warranty and Services

5 years limited

Specifications

Product Type Kit, Advanced Wind Experiment, Nacelle
For Use With (Application) Students build own wind turbine and experiment with wind power
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.