Learn More
Vernier KidWind Dowels
SDPSCI_ED
Catalog No. S04811ND Shop All Vernier Software Products
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04811ND 25 Pk.
S04810ND 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04811ND Supplier Vernier Supplier No. KWD25
Add to Cart
Add to Cart
Used for designing and testing wind turbine blades and fit into Wind Turbine Hub
  • Recommended for college, high school, and middle school science
  • Students can design blades using materials like balsa wood or chipboard sheets, and attach their blades to 0.25 in. dowels using glue or tapes
  • Dia. x L: 0.63 x 12.7cm (0.25 x 5 in.)
Specifications
Product Type KidWind Dowels
For Use With (Application) Designing and testing wind turbine blades
Quantity 25 Pk.
WARNING: Cancer - www.P65Warnings.ca.gov