Description
- IIncludes four gears with sizes 64, 32, 16 and 8 teeth
- Included hex locks secures to 9/64 in. hex-driveshaft, also used to change gears
- 8-tooth gear fits onto 2mm driveshafts on DC generators
- Recommended for College and High School
Warranty and Services
5-year limited warranty
Specifications
Specifications
| Product Type | Gear Set with Spool |
| For Use With (Application) | To increase generator RPM in experiments |
| Includes | Hex lock, gears of sizes: 64, 32, 16 and 8 teeth; Small 8-tooth gear fits on 2mm driveshafts |