Learn More
Vernier KidWind Red Blade Set
SDPSCI_ED
Catalog No. s04822nd Shop All Vernier Software Products
Change view
Click to view available options
:
Red blade set
Catalog No.
S04822ND Red blade set
1 options
Catalog No. S04822ND Supplier Vernier Supplier No. KWRED
Only null left
Add to Cart
Add to Cart
Optimized to spin at very high RPMs.
  • Produce high voltage of 5 to 6V with Wind Turbine Generator
  • 8 in. diameter fits onto any DC generator or motor with 2mm drive shaft
  • With airfoil profile and twisted pitch similar to real turbine blades
  • Used to light bulbs and pump water with basic box fan winds
  • Included with MINI Wind Turbine
  • Recommended for College, High School, and Middle School Science
Warranty and Services
5-year limited warranty
Specifications
Product Type Red Blade Set
WARNING: Cancer - www.P65Warnings.ca.gov