Learn More
Vernier KidWind Sound and Light Board
SDPSCI_ED
Catalog No. S04826ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Light and Sound board
Catalog No. Product Type
S04826ND Light and Sound board
1 options
Catalog No. S04826ND Supplier Vernier Supplier No. KWSLB
Add to Cart
Add to Cart
To demonstrate power output from small electrical devices.
  • Included in MINI wind turbine
  • Plays music or lights LED bulbs with electrical output from solar panel or small wind turbine
  • Requires 2V for functioning
  • Recommended for College, High School, and Middle School Science
Warranty and Services
5-year limited warranty
Specifications
Product Type Light and Sound board