Learn More
Vernier Microstirrer
SDPSCI_ED
Catalog No. S16012ND Shop All Vernier Software Products
Change view
Click to view available options
Type:
Microstirrer
Catalog No. Type
S16012ND Microstirrer
1 options
Catalog No. S16012ND Supplier Vernier Supplier No. MSTIR
Add to Cart
Add to Cart
For use with Conductivity Probe, pH Sensor, Tris-Compatible Flat pH Sensor, Drop Counter, and Stir Station.
  • Microstirrer takes place of stirring bar that is typically used with laboratory magnetic stirrer
Specifications
Type Microstirrer